accessibility menu, dialog, popup

UserName

NBrClCl

CAS RN 866755-20-6

IUPAC Name: 3-bromo-2,6-dichloropyridine
Synonyms: 3-bromo-2,6-dichloropyridine
Molecular Weight (g/mol): 226.88
Molecular Formula: C5H2BrCl2N
InChi Key: PZHASTRIYXMWKM-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results