CAS RN 866-97-7
CAS RN 866-97-7
IUPAC Name:
tetrapentylazanium bromide
Synonyms:
tetrapentylammonium bromide
Molecular Weight (g/mol):
378.48
Molecular Formula:
C20H44BrN
InChi Key:
SPALIFXDWQTXKS-UHFFFAOYSA-M
SMILES:
[Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
1
–
1
of
1
results
1
Tetrapentylammonium bromide, 99+%
CAS: 866-97-7 Molecular Formula: C20H44BrN Molecular Weight (g/mol): 378.48 MDL Number: MFCD00011856 InChI Key: SPALIFXDWQTXKS-UHFFFAOYSA-M Synonym: tetrapentylammonium bromide,tetraamylammonium bromide,tetra-n-amylammonium bromide,1-pentanaminium, n,n,n-tripentyl-, bromide 1:1,tetrapentylazanium bromide,n,n,n-tripentylpentan-1-aminium bromide,tetraamylammoniumbromide,1-pentanaminium, n,n,n-tripentyl-, bromide,acmc-209qb0,tetrapentyl ammonium bromide PubChem CID: 70086 SMILES: [Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC