accessibility menu, dialog, popup

UserName

CAS RN 86479-06-3

FFFFOClNHONHOFFCl

CAS RN 86479-06-3

IUPAC Name: 3-[3,5-dichloro-4-(1,1,2,2-tetrafluoroethoxy)phenyl]-1-(2,6-difluorobenzoyl)urea
Synonyms: sonet hexafluoron recruit epa pesticide chemical code 118202 hexafluron hexaflumuron sentricon
Molecular Weight (g/mol): 461.14
Molecular Formula: C16H8Cl2F6N2O3
InChi Key: RGNPBRKPHBKNKX-UHFFFAOYSA-N
SMILES: FC(F)C(F)(F)OC1=C(Cl)C=C(NC(=O)NC(=O)C2=C(F)C=CC=C2F)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results