CAS RN 86426-17-7
CAS RN 86426-17-7
IUPAC Name:
3-(2,2,2-trimethylhydrazin-2-ium-1-yl)propanoate dihydrate
Synonyms:
meldonium
Molecular Weight (g/mol):
182.22
Molecular Formula:
C6H18N2O4
InChi Key:
JFWLFLLRLZSBRA-UHFFFAOYSA-N
SMILES:
O.O.C[N+](C)(C)NCCC([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Mildronate dihydrate
CAS: 86426-17-7 Molecular Formula: C6 H14 N2 O2 . 2 H2 O