accessibility menu, dialog, popup

UserName

FFFFFFOOH

CAS RN 86301-98-6

IUPAC Name: 3-(1,1,2,3,3,3-hexafluoropropyl)adamantane-1-carboxylic acid
Synonyms: 3-(1,1,2,3,3,3-hexafluoropropyl)adamantane-1-carboxylic acid
Molecular Weight (g/mol): 330.27
Molecular Formula: C14H16F6O2
InChi Key: GLJDZZQQPFAEPQ-UHFFFAOYNA-N
SMILES: OC(=O)C12CC3CC(C1)CC(C3)(C2)C(F)(F)C(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results