CAS RN 862121-57-1
CAS RN 862121-57-1
IUPAC Name:
strontium(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms:
strontium(2+) bis(bistriflylimide anion)
Molecular Weight (g/mol):
647.89
Molecular Formula:
C4F12N2O8S4Sr
InChi Key:
DIMYICIXZTVMOY-UHFFFAOYSA-N
SMILES:
[Sr++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
Strontium bis(trifluoromethylsulfonyl)imide, Thermo Scientific™
CAS: 862121-57-1 Molecular Formula: C4F12N2O8S4Sr Molecular Weight (g/mol): 647.891 MDL Number: MFCD23380176 InChI Key: DIMYICIXZTVMOY-UHFFFAOYSA-N Synonym: Bis(trifluoromethane)sulfonimide strontium salt; Bis(trifluoromethanesulfonyl)imide strontium salt PubChem CID: 134159298 IUPAC Name: strontium;bis(trifluoromethylsulfonyl)azanide SMILES: C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sr+2]