accessibility menu, dialog, popup

UserName

CAS RN 859502-43-5

H3COHNFOONHNSOOCH3NH2SOOHO

CAS RN 859502-43-5

IUPAC Name: [1-(2-methanesulfonamidoethyl)piperidin-4-yl]methyl 5-fluoro-2-methoxy-1H-indole-3-carboxylate; sulfamic acid
Synonyms: [1-(2-methanesulfonamidoethyl)piperidin-4-yl]methyl 5-fluoro-2-methoxy-1H-indole-3-carboxylate; sulfamic acid
Molecular Weight (g/mol): 524.58
Molecular Formula: C19H29FN4O8S2
InChi Key: VQJFDBXITIOGJM-UHFFFAOYSA-N
SMILES: NS(O)(=O)=O.COC1=C(C(=O)OCC2CCN(CCNS(C)(=O)=O)CC2)C2=CC(F)=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results