CAS RN 85319-73-9
CAS RN 85319-73-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
5,6,8,9,10,11-Hexahydro-11-hydroxy-benz[a]anthracene-11-acetic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5,6,8,9,10,11-Hexahydro-11-hydroxy-benz[a]anthracene-11-acetic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5,6,8,9,10,11-Hexahydro-11-hydroxy-benz[a]anthracene-11-acetic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.