accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNO

CAS RN 852227-95-3

IUPAC Name: 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Synonyms: 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Molecular Weight (g/mol): 289.18
Molecular Formula: C16H24BNO3
InChi Key: NCJDKFFODGZRRL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results