accessibility menu, dialog, popup

UserName

CAS RN 850568-02-4

BFNH2HOOHHCl

CAS RN 850568-02-4

IUPAC Name: [2-(aminomethyl)-4-fluorophenyl]boronic acid hydrochloride
Synonyms: 2-(aminomethyl)-4-fluorophenylboronic acid hydrochloride
Molecular Weight (g/mol): 205.42
Molecular Formula: C7H10BClFNO2
InChi Key: IWBXNXDYMQDTHO-UHFFFAOYSA-N
SMILES: Cl.NCC1=C(C=CC(F)=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results