accessibility menu, dialog, popup

UserName

CAS RN 850140-72-6

IUPAC Name: N-{4-[(3-chloro-4-fluorophenyl)amino]-7-(oxolan-3-yloxy)quinazolin-6-yl}-4-(dimethylamino)but-2-enamide
Synonyms: afatinib
Molecular Weight (g/mol): 485.94
Molecular Formula: C24H25ClFN5O3
InChi Key: ULXXDDBFHOBEHA-UHFFFAOYNA-N
SMILES: CN(C)CC=CC(=O)NC1=C(OC2CCOC2)C=C2N=CN=C(NC3=CC(Cl)=C(F)C=C3)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results