accessibility menu, dialog, popup

UserName

CAS RN 844-51-9

OO

CAS RN 844-51-9

IUPAC Name: 2,5-diphenylcyclohexa-2,5-diene-1,4-dione
Synonyms: 2,5-diphenylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 260.29
Molecular Formula: C18H12O2
InChi Key: QYXHDJJYVDLECA-UHFFFAOYSA-N
SMILES: O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results