CAS RN 84-85-5
CAS RN 84-85-5
IUPAC Name:
4-methoxynaphthalen-1-ol
Synonyms:
4-methoxynaphthalen-1-ol
Molecular Weight (g/mol):
174.2
Molecular Formula:
C11H10O2
InChi Key:
BOTGCZBEERTTDQ-UHFFFAOYSA-N
SMILES:
COC1=C2C=CC=CC2=C(O)C=C1
1
–
3
of
3
results
1
4-Methoxynaphthalen-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Methoxynaphthalen-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Methoxynaphthalen-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.