accessibility menu, dialog, popup

UserName

CAS RN 82967-94-0

ClClSOONH2

CAS RN 82967-94-0

IUPAC Name: 2,3-dichlorobenzene-1-sulfonamide
Synonyms: 2,3-dichlorobenzenesulfonamide
Molecular Weight (g/mol): 226.07
Molecular Formula: C6H5Cl2NO2S
InChi Key: QJARBNAXWFCCKX-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=C(Cl)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results