CAS RN 829-85-6
CAS RN 829-85-6
IUPAC Name:
diphenylphosphane
Synonyms:
diphenylphosphine
Molecular Weight (g/mol):
186.19
Molecular Formula:
C12H11P
InChi Key:
GPAYUJZHTULNBE-UHFFFAOYSA-N
SMILES:
P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
2
of
2
results
1
Promotions Available
Diphenylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals
Diphenylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals
2
Promotions Available
Diphenylphosphine, 95%, Thermo Scientific Chemicals
Diphenylphosphine, 95%, Thermo Scientific Chemicals
CAS: 829-85-6 Molecular Weight (g/mol): 186.19 MDL Number: MFCD00003040 InChI Key: GPAYUJZHTULNBE-UHFFFAOYSA-N IUPAC Name: diphenylphosphane SMILES: P(C1=CC=CC=C1)C1=CC=CC=C1