CAS RN 82832-73-3
CAS RN 82832-73-3
IUPAC Name:
4'-propyl-[1,1'-bi(cyclohexane)]-4-one
Synonyms:
4'-propyl-[1,1'-bi(cyclohexane)]-4-one
Molecular Weight (g/mol):
222.37
Molecular Formula:
C15H26O
InChi Key:
AKCZQKBKWXBJOF-UHFFFAOYSA-N
SMILES:
CCCC1CCC(CC1)C1CCC(=O)CC1
1
–
1
of
1
results
1
trans-4-(4-n-Propylcyclohexyl)cyclohexanone, 97%, Thermo Scientific™
CAS: 82832-73-3 Molecular Formula: C15H26O Molecular Weight (g/mol): 222.372 MDL Number: MFCD04038797 InChI Key: AKCZQKBKWXBJOF-UHFFFAOYSA-N Synonym: 4-trans-4-propylcyclohexyl cyclohexanone,4-4-propylcyclohexyl cyclohexanone,4-propyldicyclohexylanone,4-trans-4'-n-propylcyclohexyl-cyclohexanone,4-4-propylcyclohexyl cyclohexan-1-one,4-trans-4'-propylcyclohexyl cyclohexanone,4'-propyl-1,1'-bicyclohexyl-4-one,4'-propyl bi cyclohexyl-4-one,1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one,pubchem2509 PubChem CID: 4354185 IUPAC Name: 4-(4-propylcyclohexyl)cyclohexan-1-one SMILES: CCCC1CCC(CC1)C2CCC(=O)CC2