CAS RN 827614-69-7
IUPAC Name:
1-propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms:
1-propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol):
236.12
Molecular Formula:
C12H21BN2O2
InChi Key:
BKLGYJWLZWMIDO-UHFFFAOYSA-N
SMILES:
CCCN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1