accessibility menu, dialog, popup

UserName

CAS RN 827614-66-4

CH3H3CNNBOCH3CH3CH3H3CO

CAS RN 827614-66-4

IUPAC Name: 1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 250.15
Molecular Formula: C13H23BN2O2
InChi Key: YMEBZRNYQBODKB-UHFFFAOYSA-N
SMILES: CC(C)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results