CAS RN 82499-03-4
CAS RN 82499-03-4
IUPAC Name:
6-bromo-7H-purin-2-amine
Synonyms:
6-bromo-7H-purin-2-amine
Molecular Weight (g/mol):
214.03
Molecular Formula:
C5H4BrN5
InChi Key:
HPGBGNVPUMCKPM-UHFFFAOYSA-N
SMILES:
NC1=NC(Br)=C2NC=NC2=N1
1
–
1
of
1
results
1
2-Amino-6-bromopurine, 98%
CAS: 82499-03-4 Molecular Formula: C5H4BrN5 Molecular Weight (g/mol): 214.026 MDL Number: MFCD01321309 InChI Key: HPGBGNVPUMCKPM-UHFFFAOYSA-N Synonym: 2-amino-6-bromopurine,6-bromo-9h-purin-2-amine,1h-purin-2-amine, 6-bromo,6-bromoguanine,d2-amino-6-bromopurine,6-bromopurine-2-ylamine,purine, 2-amino-6-bromo,6-bromo-9h-purin-2-ylamine,9h-purin-2-amine,6-bromo PubChem CID: 6101139 IUPAC Name: 6-bromo-7H-purin-2-amine SMILES: C1=NC2=C(N1)C(=NC(=N2)N)Br