accessibility menu, dialog, popup

UserName

CAS RN 824-40-8

HOONO

CAS RN 824-40-8

IUPAC Name: 2-carboxypyridin-1-ium-1-olate
Synonyms: 2-carboxypyridin-1-ium-1-olate
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: FHYMLBVGNFVFBT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=[N+]1[O-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results