accessibility menu, dialog, popup

UserName

CAS RN 82308-17-6

NiIIPPPhPhPhPhPhPh

CAS RN 82308-17-6

IUPAC Name: diiodonickel; bis(triphenylphosphane)
Synonyms: nickel(II) iodide; bis(triphenylphosphine)
Molecular Weight (g/mol): 837.09
Molecular Formula: C36H30I2NiP2
InChi Key: AKXNPPNNEDIAJL-UHFFFAOYSA-L
SMILES: I[Ni]I.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results