CAS RN 823-76-7
CAS RN 823-76-7
IUPAC Name:
1-cyclohexylethan-1-one
Synonyms:
ethanone, 1-cyclohexyl-
Molecular Weight (g/mol):
126.2
Molecular Formula:
C8H14O
InChi Key:
RIFKADJTWUGDOV-UHFFFAOYSA-N
SMILES:
CC(=O)C1CCCCC1
1
–
1
of
1
results
1
Cyclohexyl methyl ketone, 95%
CAS: 823-76-7 Molecular Formula: C8H14O Molecular Weight (g/mol): 126.199 MDL Number: MFCD00040418 InChI Key: RIFKADJTWUGDOV-UHFFFAOYSA-N Synonym: 1-cyclohexylethan-1-one,cyclohexyl methyl ketone,acetylcyclohexane,ethanone, 1-cyclohexyl,cyclohexylethanone,methyl cyclohexyl ketone,1-acetylcyclohexane,cyclohexane, acetyl,acetophenone, hexahydro,ketone, cyclohexyl methyl PubChem CID: 13207 IUPAC Name: 1-cyclohexylethanone SMILES: CC(=O)C1CCCCC1