accessibility menu, dialog, popup

UserName

CAS RN 82261-42-5

NH2N

CAS RN 82261-42-5

IUPAC Name: 4-(pyridin-3-yl)aniline
Synonyms: 4-(pyridin-3-yl)aniline
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: DKFDPLVNPGJNDE-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results