accessibility menu, dialog, popup

UserName

CAS RN 821768-06-3

NNNFFFONHNFFF

CAS RN 821768-06-3

IUPAC Name: 4-[3-(trifluoromethyl)pyridin-2-yl]-N-[5-(trifluoromethyl)pyridin-2-yl]piperazine-1-carboxamide
Synonyms: 4-[3-(trifluoromethyl)pyridin-2-yl]-N-[5-(trifluoromethyl)pyridin-2-yl]piperazine-1-carboxamide
Molecular Weight (g/mol): 419.33
Molecular Formula: C17H15F6N5O
InChi Key: JFRYYGVYCWYIDQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(NC(=O)N2CCN(CC2)C2=C(C=CC=N2)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results