CAS RN 82156-58-9
CAS RN 82156-58-9
IUPAC Name:
ethyl 2-(1-benzofuran-3-yl)acetate
Synonyms:
ethyl 2-(1-benzofuran-3-yl)acetate
Molecular Weight (g/mol):
204.23
Molecular Formula:
C12H12O3
InChi Key:
LSGDCGDXQLDKGO-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=COC2=CC=CC=C12