CAS RN 82-45-1
CAS RN 82-45-1
IUPAC Name:
1-amino-9,10-dihydroanthracene-9,10-dione
Synonyms:
1-aminoanthraquinone
Molecular Weight (g/mol):
223.23
Molecular Formula:
C14H9NO2
InChi Key:
KHUFHLFHOQVFGB-UHFFFAOYSA-N
SMILES:
NC1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O