accessibility menu, dialog, popup

UserName

CAS RN 82-43-9

ClOClO

CAS RN 82-43-9

IUPAC Name: 1,8-dichloro-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,8-dichloroanthracene-9,10-dione
Molecular Weight (g/mol): 277.1
Molecular Formula: C14H6Cl2O2
InChi Key: VBQNYYXVDQUKIU-UHFFFAOYSA-N
SMILES: ClC1=CC=CC2=C1C(=O)C1=C(Cl)C=CC=C1C2=O
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results