CAS RN 815-24-7
CAS RN 815-24-7
IUPAC Name:
2,2,4,4-tetramethylpentan-3-one
Synonyms:
2,2,4,4-tetramethylpentan-3-one
Molecular Weight (g/mol):
142.24
Molecular Formula:
C9H18O
InChi Key:
UIQGEWJEWJMQSL-UHFFFAOYSA-N
SMILES:
CC(C)(C)C(=O)C(C)(C)C
1
–
2
of
2
results
1
Hexamethylacetone, 98%
CAS: 815-24-7 Molecular Formula: C9H18O Molecular Weight (g/mol): 142.242 MDL Number: MFCD00008845 InChI Key: UIQGEWJEWJMQSL-UHFFFAOYSA-N PubChem CID: 13152 IUPAC Name: 2,2,4,4-tetramethylpentan-3-one SMILES: CC(C)(C)C(=O)C(C)(C)C