accessibility menu, dialog, popup

UserName

CAS RN 81166-84-9

CH3SiH3CCH3

CAS RN 81166-84-9

IUPAC Name: (2-cyclopropylethynyl)trimethylsilane
Synonyms: (2-cyclopropylethynyl)trimethylsilane
Molecular Weight (g/mol): 138.29
Molecular Formula: C8H14Si
InChi Key: IYQSVLJDYLPMCX-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1CC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)