CAS RN 81161-17-3
CAS RN 81161-17-3
IUPAC Name:
methyl 3-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)propanoate hydrochloride
Synonyms:
esmolol hydrochloride
Molecular Weight (g/mol):
331.84
Molecular Formula:
C16H26ClNO4
InChi Key:
GEKNCWBANDDJJL-UHFFFAOYNA-N
SMILES:
Cl.COC(=O)CCC1=CC=C(OCC(O)CNC(C)C)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Esmolol hydrochloride
CAS: 81161-17-3 Molecular Formula: C16H26ClNO4 Molecular Weight (g/mol): 331.84 InChI Key: GEKNCWBANDDJJL-UHFFFAOYNA-N IUPAC Name: methyl 3-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)propanoate hydrochloride SMILES: Cl.COC(=O)CCC1=CC=C(OCC(O)CNC(C)C)C=C1