accessibility menu, dialog, popup

UserName

CAS RN 80866-80-4

HOClNOO

CAS RN 80866-80-4

IUPAC Name: (2-chloro-5-nitrophenyl)methanol
Synonyms: (2-chloro-5-nitrophenyl)methanol
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: NNLQYDLTFRXAKD-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1Cl)[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)