CAS RN 8064-90-2
CAS RN 8064-90-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TOKU-E™ Co-trimoxazole ReadyMade™ Solution
Co-trimoxazole ReadyMade™ Solution is provided as a sterile-filtered solution of Co-trimoxazole in DMSO.