accessibility menu, dialog, popup

UserName

CAS RN 80360-14-1

SOONI

CAS RN 80360-14-1

IUPAC Name: 1-(benzenesulfonyl)-3-iodo-1H-indole
Synonyms: 1-(benzenesulfonyl)-3-iodoindole
Molecular Weight (g/mol): 383.2
Molecular Formula: C14H10INO2S
InChi Key: GKYWOZYEMLEJFK-UHFFFAOYSA-N
SMILES: IC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results