CAS RN 80278-67-7
CAS RN 80278-67-7
IUPAC Name:
isoquinoline-5-carbaldehyde
Synonyms:
isoquinoline-5-carbaldehyde
Molecular Weight (g/mol):
157.17
Molecular Formula:
C10H7NO
InChi Key:
ILRSABOCKMOFGW-UHFFFAOYSA-N
SMILES:
O=CC1=C2C=CN=CC2=CC=C1
1
–
1
of
1
results
1
Isoquinoline-5-carboxaldehyde, 97%
CAS: 80278-67-7 Molecular Formula: C10H7NO Molecular Weight (g/mol): 157.172 MDL Number: MFCD03412483 InChI Key: ILRSABOCKMOFGW-UHFFFAOYSA-N Synonym: 5-isoquinolinecarboxaldehyde,isoquinoline-5-carboxaldehyde,5-formylisoquinoline,ksc447q7n,5-isoquinolinecarbaldehyde,5-isoquinolinecarboxaldehyde 6ci,9ci PubChem CID: 7016853 IUPAC Name: isoquinoline-5-carbaldehyde SMILES: C1=CC2=C(C=CN=C2)C(=C1)C=O