accessibility menu, dialog, popup

UserName

CAS RN 80213-28-1

HClCH3SOOH2N

CAS RN 80213-28-1

IUPAC Name: hydrogen 3-methanesulfonylaniline chloride
Synonyms: hydrogen 3-methanesulfonylaniline chloride
Molecular Weight (g/mol): 207.67
Molecular Formula: C7H10ClNO2S
InChi Key: ZMPXGSYLKFAGOV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CS(=O)(=O)C1=CC=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results