CAS RN 80154-34-3
CAS RN 80154-34-3
IUPAC Name:
2-{[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}benzaldehyde
Synonyms:
2-{[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}benzaldehyde
Molecular Weight (g/mol):
284.26
Molecular Formula:
C13H16O7
InChi Key:
BGOFCVIGEYGEOF-WJTVCTBASA-N
SMILES:
OC[C@@H]1O[C@H](OC2=CC=CC=C2C=O)[C@@H](O)[C@H](O)[C@H]1O
1
–
1
of
1
results
1
4-Formylphenyl beta-D-allopyranoside, 98%, Thermo Scientific Chemicals
CAS: 80154-34-3 Molecular Formula: C13H16O7 Molecular Weight (g/mol): 284.264 MDL Number: MFCD00210992 InChI Key: OLZAGZCCJJBKNZ-SYLRKERUSA-N Synonym: helicide,helicid,hilicidum,4-formylphenyl beta-d-allopyranoside,4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde,4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde,4-formylphenyl ss-d-allopyranoside,4-beta-d-allopyranosyloxy benzaldehyde,4-formylphenyl b-d-allopyranoside PubChem CID: 12896796 IUPAC Name: 4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde SMILES: C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O