accessibility menu, dialog, popup

UserName

CAS RN 80-51-3

H2NNHSOOOSOOHNNH2

CAS RN 80-51-3

IUPAC Name: 4-[4-(hydrazinesulfonyl)phenoxy]benzene-1-sulfonohydrazide
Synonyms: 4-[4-(hydrazinesulfonyl)phenoxy]benzenesulfonohydrazide
Molecular Weight (g/mol): 358.39
Molecular Formula: C12H14N4O5S2
InChi Key: NBOCQTNZUPTTEI-UHFFFAOYSA-N
SMILES: NNS(=O)(=O)C1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)NN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results