accessibility menu, dialog, popup

UserName

CAS RN 80-41-1

CH3SOOOCl

CAS RN 80-41-1

IUPAC Name: 2-chloroethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-chloroethyl p-tosylate
Molecular Weight (g/mol): 234.69
Molecular Formula: C9H11ClO3S
InChi Key: ZXNMIUJDTOMBPV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCCl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results