CAS RN 79669-50-4
CAS RN 79669-50-4
IUPAC Name:
methyl 5-bromo-2-methylbenzoate
Synonyms:
methyl 5-bromo-2-methylbenzoate
Molecular Weight (g/mol):
229.07
Molecular Formula:
C9H9BrO2
InChi Key:
FDCYLMYCHALQJR-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Br)=CC=C1C
1
–
1
of
1
results
1
Methyl 5-bromo-2-methylbenzoate, 98%
CAS: 79669-50-4 Molecular Formula: C9H9BrO2 Molecular Weight (g/mol): 229.073 MDL Number: MFCD09839555 InChI Key: FDCYLMYCHALQJR-UHFFFAOYSA-N Synonym: methyl 5-bromo-2-methyl-benzoate,methyl 5-bromo-o-toluate,methyl5-bromo-2-methylbenzoate,5-bromo-2-methyl-benzoic acid methyl ester,benzoic acid, 5-bromo-2-methyl-, methyl ester,5-bromo-2-methylbenzoic acid methyl ester,5-bromo-2-methylbenzoicacid,methyl 4-bromo-toluate,pubchem10936,acmc-1bhme PubChem CID: 316201 IUPAC Name: methyl 5-bromo-2-methylbenzoate SMILES: CC1=C(C=C(C=C1)Br)C(=O)OC