CAS RN 79645-27-5
CAS RN 79645-27-5
IUPAC Name:
4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid
Synonyms:
sulfuric acid; tobramycin
Molecular Weight (g/mol):
565.59
Molecular Formula:
C18H39N5O13S
InChi Key:
ZEUUPKVZFKBXPW-UHFFFAOYNA-N
SMILES:
OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O
1
–
1
of
1
results
1
Melford Tobramycin Sulfate
Tobramycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, leading to the misreading of mRNA and preventing the proper initiation complex formation.