accessibility menu, dialog, popup

UserName

CAS RN 79455-63-3

FClClO

CAS RN 79455-63-3

IUPAC Name: 2-chloro-6-fluorobenzoyl chloride
Synonyms: 2-chloro-6-fluorobenzoyl chloride
Molecular Weight (g/mol): 193
Molecular Formula: C7H3Cl2FO
InChi Key: GFNAJZAKJGKJCS-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Cl)=C1C(Cl)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results