accessibility menu, dialog, popup

UserName

CAS RN 79200-57-0

NH3(R)(R)OH(R)HO(S)HO

CAS RN 79200-57-0

IUPAC Name: (1R,2S,3R,4R)-2,3-dihydroxy-4-(hydroxymethyl)cyclopentan-1-aminium
Synonyms: (1R,2S,3R,4R)-2,3-dihydroxy-4-(hydroxymethyl)cyclopentan-1-aminium
Molecular Weight (g/mol): 148.18
Molecular Formula: C6H14NO3
InChi Key: FHNKBDPGQXLKRW-KAZBKCHUSA-O
SMILES: [NH3+][C@@H]1C[C@H](CO)[C@@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results