CAS RN 79200-56-9
CAS RN 79200-56-9
IUPAC Name:
(1R,4S)-2-azabicyclo[2.2.1]hept-5-en-3-one
Synonyms:
(1R,4S)-2-azabicyclo[2.2.1]hept-5-en-3-one
Molecular Weight (g/mol):
109.13
Molecular Formula:
C6H7NO
InChi Key:
DDUFYKNOXPZZIW-UHNVWZDZSA-N
SMILES:
O=C1N[C@@H]2C[C@H]1C=C2
1
–
1
of
1
results
1
(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific Chemicals
CAS: 79200-56-9 Molecular Formula: C6H7NO Molecular Weight (g/mol): 109.13 MDL Number: MFCD00211274 InChI Key: DDUFYKNOXPZZIW-UHNVWZDZSA-N Synonym: 1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one,--1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one,1s,4r-3-azabicyclo 2.2.1 hept-5-en-2-one,2-azabicyclo 2,2,1 hept-5-en-3-one,vince lactam,2-azabicyclo 2.2.1 hept-5-en-3-one, 1r,4s,--2-azabicyclo 2.2.1 hept-5-en-3-one,--vince lactam,1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one PubChem CID: 2725037 IUPAC Name: (1S,4R)-3-azabicyclo[2.2.1]hept-5-en-2-one SMILES: C1C2C=CC1NC2=O