CAS RN 791-31-1
CAS RN 791-31-1
IUPAC Name:
triphenylsilanol
Synonyms:
triphenylsilanol 1,1,1-triphenyl-silanol silane, (hydroxytriphenyl)- C18H16osi Si(OH)Ph3 (hydroxytriphenyl)-silane silanol, 1,1,1-triphenyl- silanol, triphenyl-
Molecular Weight (g/mol):
276.41
Molecular Formula:
C18H16OSi
InChi Key:
NLSXASIDNWDYMI-UHFFFAOYSA-N
SMILES:
O[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Triphenylsilanol, 98%
CAS: 791-31-1 Molecular Formula: C18H16OSi Molecular Weight (g/mol): 276.41 MDL Number: MFCD00002102 InChI Key: NLSXASIDNWDYMI-UHFFFAOYSA-N Synonym: triphenylsilanol,silanol, triphenyl,triphenylhydroxysilane,silane, hydroxytriphenyl,unii-azc8o4ttx4,silanol, 1,1,1-triphenyl,hydroxy triphenyl silane,si oh ph3,azc8o4ttx4 PubChem CID: 69925 ChEBI: CHEBI:35035 IUPAC Name: hydroxy(triphenyl)silane SMILES: C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O