accessibility menu, dialog, popup

UserName

CAS RN 79032-48-7

H3CONHOHOCH3H3CSNO

CAS RN 79032-48-7

IUPAC Name: 2-acetamido-3-methyl-3-(nitrososulfanyl)butanoic acid
Synonyms: 2-acetamido-3-methyl-3-(nitrososulfanyl)butanoic acid
Molecular Weight (g/mol): 220.24
Molecular Formula: C7H12N2O4S
InChi Key: ZIIQCSMRQKCOCT-UHFFFAOYNA-N
SMILES: CC(=O)NC(C(O)=O)C(C)(C)SN=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results