accessibility menu, dialog, popup

UserName

CH3CH3CH3CCH3CH3CH3CH3CCH3Ni

CAS RN 79019-60-6

IUPAC Name: nickel(2+) bis(2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide)
Synonyms: nickel(2+) bis(2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide)
Molecular Weight (g/mol): 301.1
Molecular Formula: C18H26Ni
InChi Key: PPBIVTWBQQUEKM-UHFFFAOYNA-N
SMILES: [Ni++].CC1[C-]=C(C)C(C)=C1C.CC1[C-]=C(C)C(C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results