accessibility menu, dialog, popup

UserName

CAS RN 787-69-9

H3COCH3O

CAS RN 787-69-9

IUPAC Name: 1-{4'-acetyl-[1,1'-biphenyl]-4-yl}ethan-1-one
Synonyms: 1-{4'-acetyl-[1,1'-biphenyl]-4-yl}ethanone
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: YSTSBXDVNKYPTR-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)