CAS RN 78617-12-6
IUPAC Name:
2-(heptyloxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
heptyl glucoside �-D-galactopyranoside, heptyl
Molecular Weight (g/mol):
278.35
Molecular Formula:
C13H26O6
InChi Key:
NIDYWHLDTIVRJT-UHFFFAOYNA-N
SMILES:
CCCCCCCOC1OC(CO)C(O)C(O)C1O
1
–
1
of
1
results
1
Heptyl beta-D-glucopyranoside, 99%
CAS: 78617-12-6 Molecular Formula: C13H26O6 Molecular Weight (g/mol): 278.345 MDL Number: MFCD00063299 InChI Key: NIDYWHLDTIVRJT-UJPOAAIJSA-N Synonym: heptyl-beta-d-glucopyranoside,heptyl beta-d-glucopyranoside,2r,3r,4s,5s,6r-2-heptyloxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol,b7g,n-heptyl beta-d-glucopyranoside,n-heptyl-i 2-d-glucopyranoside,heptyl-?-d-glucopyranoside,heptyl b-d-glucopyranoside,heptyl-b-d-glucopyranoside,d06ibv PubChem CID: 448173 IUPAC Name: (2R,3R,4S,5S,6R)-2-heptoxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: CCCCCCCOC1C(C(C(C(O1)CO)O)O)O