accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFFFFFSiClClCl

CAS RN 78560-44-8

IUPAC Name: trichloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
Synonyms: perfluorodecyltrichlorosilane
Molecular Weight (g/mol): 581.55
Molecular Formula: C10H4Cl3F17Si
InChi Key: VIFIHLXNOOCGLJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results