accessibility menu, dialog, popup

UserName

H3COFBOHHO

CAS RN 78495-63-3

IUPAC Name: (2-fluoro-6-methoxyphenyl)boronic acid
Synonyms: 2-fluoro-6-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: XOVMDVZAWWQSDC-UHFFFAOYSA-N
SMILES: COC1=CC=CC(F)=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results